Documenti
Z219924
Sigma-Aldrich
Aldrich® hollow pennyhead stopper
joint: ST/NS 19/38
Sinonimo/i:
glass stopper, glass stoppers, stoppers
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.35
Materiali
ground glass
Giunto
joint: ST/NS19/38
Descrizione
closed at bottom
Descrizione generale
Conforms to standard-taper specifications as outlined in ASTM E 675.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC