Documenti
Z215430
Rigid PTFE sleeve
joint: ST/NS 24/40
Sinonimo/i:
ptfe sleeves
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Giunto
joint: ST/NS24/40
Produttore/marchio commerciale
Cowie
Descrizione generale
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.