Documenti fondamentali
Z180211
Supelco
Bottom-drip adapter for large columns
1/4-28 UNF 2B internal thread and flow-regulator valve, for use with tubing connectors, PTFE
About This Item
Codice UNSPSC:
12000000
NACRES:
SB.52
Materiali
PTFE
Produttore/marchio commerciale
Ace Glass 583855
tecniche
LPLC: suitable
Descrizione generale
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred #50 and include one FETFE® O-ring.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)