Documenti fondamentali
Z179558
Supelco
Pre-column for Michel-Miller columns
About This Item
Codice UNSPSC:
12352200
NACRES:
SB.52
Produttore/marchio commerciale
Ace Glass 579634
tecniche
LPLC: suitable
Lungh. × D.I.
130 mm × 22 mm
Descrizione generale
For the addition of pre-purified sample mixtures to main column. Technique allows silica gel or alumina-packed columns to be reused several times. Suitable for use in final stages of slurry packing; stacks on top of main M-M column using coupling. Connections are Ace Thred #11.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)