Documenti fondamentali
Z176044
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter without return hole
female joint: ST/NS 24/40, male joint: ST/NS 24/40, capacity 100 mL
Scegli un formato
1 EA
CHF 153.00
CHF 153.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 153.00
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 153.00
Check Cart for Availability
Giunto
female joint: ST/NS24/40
male joint: ST/NS24/40
Capacità
100 mL
Categorie correlate
Descrizione generale
Aldrich® antisplash splash-guard adapter without return hole are traps that greatly reduce carry-over from foaming or bumping. Whatever comes up into this trap will stay and not go back to the evaporator flask.
Applicazioni
Aldrich® antisplash splash-guard adapter without return hole is ideal for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping".
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)