Z168645
Diaphragm-seal packless valve
1/4 turn, 316 stainless steel
About This Item
Codice UNSPSC:
40141600
NACRES:
NB.51
Materiali
316 stainless steel
Descrizione generale
1/4 in. NPTM threads. Diffusion and corrosion resistant. KEL-F valve seats for positive shut-off. Low rms internal finish. Body stamped with heat code for material traceability. For purity assurance, poly-bagged after electropolishing and ultrasonic cleaning. Low leak rates: <2 × 10-8 scc/s He. Pressure range: vacuum to 3,000 psig.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)