Documenti fondamentali
Z154350
Ace Trubore® 3-piece glass stirrer bearing
joint: ST/NS 45/50, fits shaft o.d., 10 mm
Scegli un formato
1 EA
CHF 155.00
CHF 155.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 155.00
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.72
CHF 155.00
Check Cart for Availability
Giunto
joint: ST/NS45/50
Caratteristiche
fits shaft o.d. 10 mm
Produttore/marchio commerciale
Ace Glass 8042121
Descrizione generale
Self-aligning, three-piece bearing consisting of a glass inner Trubore bearing, nylon bushing with O-ring, and threaded glass adapter. Inner bearing will spin if shaft binds or will self-align at O-ring if motor is slightly cocked. Inner bearing and bushing can be used with any joint size.
Note legali
Trubore is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)