Documenti
Z118338
Cotton blend glove liners
size S/M, light weight
Sinonimo/i:
Cotton Glove Inserts, Cotton Glove Liners, resuable fabric glove
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
Materiali
(52%Polyester/48%Cotton)
white gloves
Misura
S/M
Sterilità
non-sterile
Caratteristiche
latex free
powder-free
reusable
Confezionamento
pack of 12 pair
Produttore/marchio commerciale
Sigma-Aldrich
Lunghezza
9 in.
Descrizione generale
Use these gloves for product protection during inspection or light manufacturing. Also ideal for use as a liner underneath outer gloves. Reversible pattern allows glove to be worn on either hand. Lightweight styles feature unhemmed cuffs.