Documenti fondamentali
Z114227
Supelco
Hand-operated aluminum cap crimper
O.D. 11 mm
Scegli un formato
1 EA
CHF 630.00
CHF 630.00
Check Cart for Availability
Richiedi un ordine bulk
Scegli un formato
Cambia visualizzazione
1 EA
CHF 630.00
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
CHF 630.00
Check Cart for Availability
Richiedi un ordine bulk
D.E.
11 mm
Descrizione generale
  • Used to attach aluminum seals to bottles and vials with a crimp/serum finish
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Polished crimping jaws provide consistent sealing
  • 2 year warranty
Altre note
Includes five inserts to provide a double-aggressive liner seal when used with open-top seals.
Prodotti correlati
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Hand crimper, adjustable Hand crimper, adjustable, for use with 20 mm crimp seals, pkg of 1 ea
22316-U
Hand crimper, adjustable
Crimpers Hand crimper, for use with 20mm seals, pkg of 1 ea
33280-U
Crimpers
Vial Decapper, pliers-type for use with 20mm crimp seals, pkg of 1 ea
33283
Vial Decapper, pliers-type