Documenti
TEM-CF150NI50
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
size 150 mesh, box of 50
Sinonimo/i:
CARBON FILM 150 MESH NI 50/BX
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 50
Misura
150 mesh
Descrizione generale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 150 mesh.