SYNV200100G
Synthware™ micro Dewar vacuum trap
100 mL, stopcock: glass
Sinonimo/i:
Synthware V200100G
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware V200100G
Capacità
100 mL
Categorie correlate
Descrizione generale
Small scale dry ice, Dewar style trap. The dry-ice chamber is 40 mm I.D.× 60 mm in length. Choice PTFE stopcock for grease-free operation.
Note legali
Synthware is a trademark of Kemtech America Inc.