Documenti fondamentali
SYNS732510
Synthware sublimator, complete
10 mL, complete
Sinonimo/i:
Synthware S732510, sublimation apparatus
Scegli un formato
1 EA
CHF 271.00
CHF 271.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 271.00
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
CHF 271.00
Check Cart for Availability
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S732510
Capacità
10 mL
Descrizione generale
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed. The sublimate collects on the lower end of the condenser which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring and stainless steel pinch clamp.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)