Documenti fondamentali
SYNS680012
Synthware low hold up high vacuum valve with metering plug
size 0-12 mm
Sinonimo/i:
Synthware S680012
Scegli un formato
1 EA
CHF 121.00
CHF 121.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 121.00
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
CHF 121.00
Check Cart for Availability
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S680012
Descrizione generale
Viton® O-ring on PTFE shaft are protected with precision PTFE Wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal. This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals. With two 9mm O.D. sidearm bent to minimize hold up of liquids.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)