Documenti fondamentali
SYNM411004
Synthware all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left-right, Rear-left-right
Sinonimo/i:
Synthware M411004, vacuum manifold, vacuum unit
Scegli un formato
1 EA
CHF 726.00
CHF 726.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 726.00
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
CHF 726.00
Check Cart for Availability
Caratteristiche
number of ports 4
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware M411004
Descrizione generale
With 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mm stopcocks is 75 mm center to center.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)