Documenti fondamentali
SYNM260004
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left-right, Rear-left
Sinonimo/i:
Synthware M260004, vacuum manifold, vacuum unit
Scegli un formato
1 EA
CHF 839.00
CHF 839.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 839.00
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
CHF 839.00
Check Cart for Availability
Caratteristiche
number of ports 4
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware M260004
Descrizione generale
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)