Documenti fondamentali
SYNM101003
Synthware Vacuum/Inert Gas Manifold with double bank and high vacuum valves
number of ports, 3, Hose Connections: Front-left, Rear-right
Sinonimo/i:
Synthware M101003, vacuum manifold, vacuum unit
Scegli un formato
1 EA
CHF 533.00
CHF 533.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 533.00
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
CHF 533.00
Check Cart for Availability
Caratteristiche
number of ports 3
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware M101003
Descrizione generale
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm. With front-left and rear-right 10 mm hose connections.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)