Documenti fondamentali
SYND222434
Synthware Abderhalden drying chamber
top joint: ST/NS 24/40, bottom joint: ST/NS 24/40, chamber side joint: ST/NS 34/45
Sinonimo/i:
Synthware D222434
Scegli un formato
1 EA
CHF 188.00
CHF 188.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 188.00
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
CHF 188.00
Check Cart for Availability
Giunto
bottom joint: ST/NS24/40
side joint: ST/NS chamber34/45
top joint: ST/NS24/40
Produttore/marchio commerciale
D222434
Categorie correlate
Descrizione generale
Synthware Abderhalden drying chamber is designed for drying samples for elemental analysis. The sample is placed in a vial, which is then placed into the drying chamber. It operates under vacuum with gentle heat created by refluxing low boiling point solvents in the outer jacket.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)