SYNA254024
Synthware™ enlarging adapter
top joint: ST/NS 40/50, bottom joint: ST/NS 24/40
Sinonimo/i:
Synthware A254024, glass connecting adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
bottom joint: ST/NS24/40
joint: ST/NS top40/50
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A254024
Descrizione generale
Synthware™ enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (40/50) is larger that the bottom joint (24/40).
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)