Documenti fondamentali
SLW4507/45D
Pyrex® Membrane screwcap, 0.2 um PTFE membrane, GL 45 screwthread
Sinonimo/i:
Pyrex, SciLabware 4507/45D
Scegli un formato
5 EA
CHF 46.70
CHF 46.70
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
5 EA
CHF 46.70
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 46.70
Check Cart for Availability
Materiali
PTFE membrane
polypropylene screw cap
Produttore/marchio commerciale
4507/45D
SciLabware
Dimens. membrana
0.2 μm
Raccordi
GL 45
Descrizione generale
  • Polypropylene screwcap with integral 0.2 um PTFE membrane
  • Gas permeable membrane permits pressure equalisation allowing glassware to be sterilised using an autoclave with the screwcap fastened
  • Provides a tight seal that prevents the ingress of liquids or solids
  • Ideal for aerobic microbiology and cell culture
  • Suitable for storage and transport of gas generating media
  • Autoclavable
  • Temperature resistant to 140 deg. C
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)