Documenti fondamentali
SLW4503/22
Pyrex® Culture tube caps with insert
thread size 22 mm
Sinonimo/i:
Pyrex, SciLabware 4503/22
Scegli un formato
20 EA
CHF 82.30
CHF 82.30
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
20 EA
CHF 82.30
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
CHF 82.30
Check Cart for Availability
Misura filettatura
22 mm
Produttore/marchio commerciale
4503/22
SciLabware
Descrizione generale
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used.
Caratteristiche e vantaggi
  • Manufactured from borosilicate glass 3.3
  • High resistance to attack from water reduces leaching of contaminants which can cause pH changes
  • PBT screwcaps with inert PTFE lined rubber disc
  • Cap can withstand repeated dry heat and steam sterilisation cycles.
Altre note
Cap and insert only.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)