Documenti fondamentali
PL800GB
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 9 (for 8 in. ports)
About This Item
Codice UNSPSC:
46181504
NACRES:
HA.03
Materiali
butyl rubber
Misura
9 (for 8 in. ports)
Caratteristiche
reusable
Confezionamento
pack of 1 pair
Produttore/marchio commerciale
Plas-Labs 800GB
Descrizione generale
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)