Documenti fondamentali
PL800GANIT-10
Plas-Labs replacement nitrile gloves
for 2 piece glove set, size 10 (for 8 in. ports)
Scegli un formato
1 PAIR
CHF 117.00
CHF 117.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 PAIR
CHF 117.00
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
CHF 117.00
Check Cart for Availability
Materiali
nitrile
Misura
10 (for 8 in. ports)
Caratteristiche
reusable
for use with glove box sleeves
Confezionamento
pack of 1 pair
Produttore/marchio commerciale
Plas-Labs 800GANIT-10
Descrizione generale
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
Note legali
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)