MT51107874
Mettler Toledo DX series ISE half-cell membrane kit
For Lead ISE
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
Produttore/marchio commerciale
Mettler-Toledo 51107874
Descrizione generale
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE!
Kit contains one membrane plus electrolyte
Kit contains one membrane plus electrolyte
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)