Documenti fondamentali
MT51107681
Mettler Toledo DX series ISE half-cells electrode
Cyanide selective
Scegli un formato
1 EA
CHF 1’550.00
CHF 1’550.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 1’550.00
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
CHF 1’550.00
Check Cart for Availability
Materiali
membrane (Solid state)
shaft (POM)
Produttore/marchio commerciale
Mettler-Toledo 51107681
Campo di misura
1.0-1.0-3
Campo di misura della temperatura
0-80 °C
pH
4-13
Descrizione generale
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your ISE half-cell, and you have a new ISE!
electrolyte for reference electrode: 1 mol/L KNO3, ISA solution: 1 mol/L KNO3
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)