Documenti
HAM59000-40
Hamilton® Microlab® 600 instrument syringe
model, 1002 BFP, volume 2.5 mL
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Materiali
Gastight Borosilicate glass/PTFE syringe
Caratteristiche
model 1002 BFP
Produttore/marchio commerciale
Hamilton 59000-40
Parametri
10-115 °C temp. range (50-239 °F)
200 psig pressure
Diam. int. × diam. est.
7.29 mm × 10.3 mm
Volume
2.5 mL
Categorie correlate
Note legali
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.