Documenti
GF44257248
Phosphor bronze alloy, Cu94Sn6
sheet, 300 x 300mm, 3.2mm thickness, hard
Sinonimo/i:
CU053150
About This Item
Codice UNSPSC:
11101711
NACRES:
NA.23
Forma fisica
wire
Produttore/marchio commerciale
Goodfellow 442-572-48
Misura × spessore
300x300 mm × 3.2 mm