Documenti fondamentali
ESYNTH010
Sigma-Aldrich
SynLectro Stainless Steel Electrode
size 3 mm × 20 mm × 60 mm
Scegli un formato
1 EACH
CHF 111.00
CHF 111.00
Spedizione prevista il29 marzo 2025
Richiedi un ordine bulk
Scegli un formato
Cambia visualizzazione
1 EACH
CHF 111.00
About This Item
Codice UNSPSC:
81141502
NACRES:
NB.61
CHF 111.00
Spedizione prevista il29 marzo 2025
Richiedi un ordine bulk
Misura
3 mm × 20 mm × 60 mm
Descrizione generale
SynLectro Stainless Steel Electrode is a 3x20x60 mm electrode designed for use with the SynLectro Electrolysis Platform for synthetic electrochemistry.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Note legali
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
Contenuto correlato
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Questions
  1. What is the grade of your SynLectro Stainless Steel Electrodes?
    1 answer
    1. These electrodes are grade 304 stainless steel. Grade 304 stainless steel is generally regarded as the most common austenitic stainless steel. This grade contains a high nickel content, typically between 8 and 10.5 percent by weight, and a high amount of chromium, approximately 18 to 20 percent by weight. The high amounts of chromium and nickel give 304 stainless steel excellent corrosion resistance.