Documenti fondamentali
DWK73802-18400
KIMBLE® Solid Cap
18-400 thread, black phenolic cap (with rubber liner)
Scegli un formato
1 EACH
CHF 183.00
CHF 183.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EACH
CHF 183.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
CHF 183.00
Check Cart for Availability
Materiali
PTFE/rubber liner
black phenolic cap (with rubber liner)
Sterilità
non-sterile
Caratteristiche
class A
Confezionamento
case of 144 ea
Produttore/marchio commerciale
Kimble® 73802-18400
Raccordi
18-400 thread
Descrizione generale
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
Identified in ASTM Specification E982, Class A requirement
Caratteristiche e vantaggi
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)