Documenti fondamentali
DWK62121D-10
Kimble® 33 Expansion Tubular Borosilicate Glass Serum Vial Without Closure
Scegli un formato
864 EACH
CHF 1’180.00
CHF 1’180.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
864 EACH
CHF 1’180.00
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.43
CHF 1’180.00
Check Cart for Availability
Materiali
Borosilicate glass/PTFE tube
tubulated bottom
Caratteristiche
closure type: no
Confezionamento
pkg of 864 ea
Produttore/marchio commerciale
Kimble® 62121D-10
Diam. tubo
20 mm
Descrizione generale
Autoclavable serum vials are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, culture media, chromatography samples, and more.
Applicazioni
Storage and transport of samples, lyophilization, contain harsh reagents or liquids.
Caratteristiche e vantaggi
  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Aluminum seals, stoppers and septa are available separately
  • Converted from Type I borosilicate glass tubing, vials conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Questions
  1. Qué cápsula con septum podemos incorporar a la referencia DWK62121D-10? Gracias, Esther..
    1 answer
    1. Helpful?