Documenti fondamentali
DWK218067358
DURAN® graduated amber laboratory bottle
with cap, round amber glass bottle, bottle capacity (5,000 mL), non-sterile
Sinonimo/i:
218067358
Scegli un formato
1 EACH
CHF 288.00
CHF 288.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EACH
CHF 288.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 288.00
Check Cart for Availability
Materiali
round amber glass bottle
Sterilità
non-sterile
Caratteristiche
with cap
Confezionamento
case of 1 ea
Produttore/marchio commerciale
DURAN® 218067358
Capacità bottiglia
(5,000 mL)
Descrizione generale
Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties.
Very durable and chemically resistant amber colour. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print. Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)