Documenti fondamentali
DWK218066353
DURAN® graduated amber laboratory bottle
with cap, round amber glass bottle, bottle capacity (2,000 mL), non-sterile
Sinonimo/i:
218066353, DURAN Amber Bottle
Scegli un formato
10 EACH
CHF 1’010.00
CHF 1’010.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EACH
CHF 1’010.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 1’010.00
Check Cart for Availability
Materiali
round amber glass bottle
Sterilità
non-sterile
Caratteristiche
with cap
Confezionamento
case of 10 ea
Produttore/marchio commerciale
DURAN® 218066353
Capacità bottiglia
(2,000 mL)
Descrizione generale
Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties.
Very durable and chemically resistant amber colour. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print. Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Caratteristiche e vantaggi
  • Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
  • The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties
  • Very durable and chemically resistant amber colour
  • Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
  • Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)