Documenti fondamentali
CLS640260
Pyrex® Squibb separatory funnel, Pyrex jointed stopper, PTFE stopcock
Sinonimo/i:
Corning® 6402
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Misura rubinetto
2
Misura tappo a cono
16
Produttore/marchio commerciale
Corning 6402-60
D.E. stelo × lungh.
10 mm × 65 mm
Altre note
Designed so that only PTFE and borosilicate glass are in contact with liquids or gases. Two sealing mechanisms are utilized. First, the tip of the PTFE body is "threaded down" into a glass seat. Second, an adjustable gland seal within PTFE body prevents leakage through the cap. No lubrication, O-rings, or retaining clips required.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)