CLS4315100
Pyrex® pear-shaped flask, with ST 14/20 outer joint
capacity 100 mL
Sinonimo/i:
Corning® 4315
About This Item
Prodotti consigliati
Materiali
borosilicate glass
pear-shaped flask
short
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper joint
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 4315-100
Capacità
100 mL
Dimens. collo
(1)
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.