Documenti fondamentali
BR88655
BRAND® stopcock T-bore with nozzles
for tube I.D. 4 mm
Sinonimo/i:
PTFE, Stopcocks
Scegli un formato
1 EA
CHF 58.00
CHF 58.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 58.00
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
CHF 58.00
Check Cart for Availability
Materiali
PTFE
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 88655
Diam. int.
2 mm
Categorie correlate
Descrizione generale
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)