Merck
BR782881
BRAND® BRANDplates® for inserts
6-well with lid cellGrade™ S
Sinonimo/i:
BRANDplates®, Cell culture, Insert System, Inserts, Plates, Tissue culture
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Informazioni su questo articolo
NACRES:
NB.43
UNSPSC Code:
41121800
Material:
polystyrene , transparent wells
Size:
6 well
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
polystyrene , transparent wells
sterility
sterile
feature
lid, single packed
manufacturer/tradename
BRAND 782881
packaging
pack of 10 ea, pkg of Individually wrapped, with lid
size
6 well
suitability
suitable for (bacteriological assays)
General description
PS. pureGrade S surface, untreated, sterile, cover with condensation rings.Manufactured according to the requirements in ANSI/SLAS Standards 1 and 4.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Features and Benefits
  • Non-treated surface
  • Side well access for easier pipetting and removing cover slips
  • Perfect positioning of the BRANDplates® insert
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® BRANDplates® for inserts
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)