BR708745
BRAND® disposable BLAUBRAND® micropipettes, intraMark
tube capacity 50-100 μL , Dual marks, blue
Sinonimo/i:
Micropipettes, Pipettes, intraMARK
About This Item
Prodotti consigliati
Materiali
blue
Caratteristiche
DE-M marking, To Contain
Confezionamento
pack of 1000 ea (4 boxes of 250)
Produttore/marchio commerciale
BRAND 708745
Lunghezza
125 mm
Capacità
50-100 μL , Dual marks
Categorie correlate
Descrizione generale
DIN ISO 7550. DE-M marking for volumes greater than 5 µl. Calibrated to contain (TC, In). Color code to ISO for clear identification.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG