Documenti fondamentali
BR62010
BRAND® PFA sample jar with screw cap
capacity 60 mL
Sinonimo/i:
Jars, PFA
About This Item
Codice UNSPSC:
41121800
Materiali
PFA
Produttore/marchio commerciale
BRAND 62010
Capacità
60 mL
Raccordi
GL 40
Descrizione generale
For sample collection, transport and storage. PFA screw cap with molded sealing ring for a positive seal. Autoclavable, easy to clean, withstand temperatures from -200 °C to 260 °C.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)