Documenti fondamentali
BR578000
BRAND® burette clamp, aluminum die casting for 2 burettes, with roll mounting
Sinonimo/i:
Accessories, Burette clamp, Burettes
Scegli un formato
1 EA
CHF 81.00
CHF 81.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 81.00
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.43
CHF 81.00
Check Cart for Availability
Produttore/marchio commerciale
BRAND 78000
Categorie correlate
Descrizione generale
Burettes up to 50 ml can be mounted in seconds. PVC-coated clamps provide a secure grip. Comfortable height adjustment. Graduations remain visible. Very sturdy design, easy to operate. Integral support connector.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)