Documenti fondamentali
BR130422
BRAND® square bottle with tamper-proof screw cap, PE-HD
capacity 1000 mL, wide-neck
Sinonimo/i:
Bottles, Plastics, Square bottles
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
high-density polyethylene bottle
polypropylene cap
square bottle
translucent bottle
wide-mouth bottle
wide-neck
Giunto
Joints threaded neck (GL54)
Caratteristiche
with cap
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 130422
Lungh. × Alt.
85 mm × 210 mm
Capacità
1000 mL
Descrizione generale
Translucent. Screw cap with sealing cone, PP, blue, with cadmium-free pigments. Tamper-proof closure with press-off retaining ring. Space-saving square base.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)