Documenti
BR114130
BRAND® culture tube, rimless
L × W 18 mm × 180 mm, thickness 0.8 mm
Sinonimo/i:
Culture tubes, Tubes
About This Item
Codice UNSPSC:
41121706
NACRES:
NB.22
Materiali
round bottom
screw top cap
soda-lime glass tube
straight neck
transparent tube
Sterilità
non-sterile
Caratteristiche
RCF 900 × g
neck thread push fit
Confezionamento
pack of 121 ea
Produttore/marchio commerciale
BRAND 114130
Lungh. × largh.
18 mm × 180 mm
Capacità
18 mL
30 mL
Spessore
0.8 mm
Descrizione generale
Crafted from high-quality glass, these tubes provide excellent clarity and durability, making them ideal for cell culture and other scientific applications. Autoclavable at 121 °C, ensuring easy sterilization.
Caratteristiche e vantaggi
• Excellent sample visibility• Good chemical resistance
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG