Documenti fondamentali
BAH189620000
SP Bel-Art® Poxygrid® Microliter Pipettor Rack
to hold, 10 pipettors
Sinonimo/i:
Pipette Rack
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Prezzi e disponibilità al momento non sono disponibili
Caratteristiche
to hold 10 pipettors
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H18962-0000
Lungh. × Largh. × Alt.
9 in. (229 mm) × 5 in. (127 mm) × 6.75 in. (171 mm)
Descrizione generale
Store pipettors while protecting pipettor tips in these epoxy-coated aluminum racks that provide quick, easy access for more efficient work.
Caratteristiche e vantaggi
  • Holds up to 10 round shape pipettors such as Finnpipette®, Socorex, Eppendorf® and other popular brands; two end holes fit Pipette-Aid pipette fillers
  • Two stepped rows of 5 each
  • Second lower support row has smaller holes to accept tapered ends of the pipettors to keep them vertical
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Eppendorf is a registered trademark of Eppendorf AG
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)