Documenti fondamentali
BAF249760002
Workshields for β-emitters, free standing
W × L × H 30.5 cm (12 in.) × 35.6 cm (14 in.) × 52.7 cm (20.75 in.), size M, pack of 1 ea
About This Item
Codice UNSPSC:
26142300
NACRES:
NB.35
Misura
M
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F24976-0002
Largh. × lungh. × alt.
30.5 cm (12 in.) × 35.6 cm (14 in.) × 52.7 cm (20.75 in.)
Descrizione generale
Select shield size to match requirements.
  • 9.5 mm (3/8 in.) thick, glass transparent acrylic shields protect against Beta sources and splashes from lab procedures
  • One piece construction has rounded corner for easy cleaning
  • Non skid feet prevent slipping on benchtop
  • Freestanding design has an angled shield and a flat base with a large work surface
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)