Documenti fondamentali
BAF203201660
SP Bel-Art® Safe-Lab Joint Stirrer Bearing
for 24/40 Tapered Joints
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Prezzi e disponibilità al momento non sono disponibili
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F20320-1660
Descrizione generale
For use with 10mm stirring rods, the body is PTFE with a Viton® O-ring.
Caratteristiche e vantaggi
  • Comes complete with two PTFE inserts with inside O-ring for fine adjustment of stirring rod tension
  • Threaded polypropylene locking compression cap on top
  • Extracting nut above the ground joint allows easy and quick removal from flask
  • Not autoclavable
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)