Documenti fondamentali
924601
Sigma-Aldrich
Cross-shaped channel chip (slide format)
Fluidic 160, PMMA with Mini-Luer Interface
Scegli un formato
1 EACH
CHF 94.30
CHF 94.30
Spedizione prevista il20 maggio 2025
Richiedi un ordine bulk
Scegli un formato
Cambia visualizzazione
1 EACH
CHF 94.30
About This Item
Codice UNSPSC:
42142600
NACRES:
NA.23
CHF 94.30
Spedizione prevista il20 maggio 2025
Richiedi un ordine bulk
Applicazioni
This is a Cross-shaped channel chip in a microscopy slide format. These chips offer two separate channel structures with crossing channels on each device. One of those with, one without a channel offset. The microfluidic chips of this family feature Mini Luer interfaces.
Cross-shaped channel chip (slide format), Fluidic 160, is made of polymethyl methacrylate (PMMA) with Mini-Luer Interface.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Recent advances of controlled drug delivery using microfluidic platforms.
Sanjay, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2020)
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired
Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range