Merck
Accessory gene regulator (agr) dysfunction in Staphylococcus aureus bloodstream isolates from South Korean patients.
Antimicrobial agents and chemotherapy (2012-12-21)
Yong Pil Chong, Eun Sil Kim, Su-Jin Park, Ki-Ho Park, Tark Kim, Mi-Na Kim, Sung-Han Kim, Sang-Oh Lee, Sang-Ho Choi, Jun Hee Woo, Jin-Yong Jeong, Yang Soo Kim
RÉSUMÉ
We describe the genetic and microbiological characteristics of methicillin-resistant Staphylococcus aureus (MRSA) bloodstream isolates with agr dysfunction from a tertiary-care hospital in Korea. Of these, ST5-SCCmec type II-agr group II MRSA isolates, which are known to be prevalent in hospital-acquired infections in Korea, were the most abundant, because of the clonal spread of a specific agr-defective lineage. This finding suggests that the loss of agr function may confer a potential advantage in a hospital setting. Clonal spread of a specific defective-agr strain was not observed among community-associated MRSA or methicillin-susceptible S. aureus clones, regardless of community or hospital acquisition of infection. agr-defective clones, including ST5 and ST239 MRSA, were enriched for heteroresistant vancomycin-intermediate S. aureus.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Methicillin sodium salt, ≥95% (HPLC)
Millipore
MRSA Selective Supplement, suitable for microbiology