- [Chemical constituents of Adenophora stricta Miq].
[Chemical constituents of Adenophora stricta Miq].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1990-08-01)
P Jiang, Z Gao
PMID2093322
RÉSUMÉ
Four compounds were isolated from the roots of Adenophora stricta. On the basis of spectrometric analysis and physicochemical constants, they were identified as beta-sitosterol, beta-sitosterol-O-beta-D-glucopyranoside, taraxerone and octacasanoic acid respectively. Taraxerone and octacasanoic acid were isolated from the plants of Adenophora for the first time.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Le tarif et la disponibilité ne sont pas disponibles actuellement.