Merck
Purification and characterization of a novel endo-β-1,4-glucanases , AfEG22, from the giant snail, Achatina fulica frussac.
Acta biochimica et biophysica Sinica (2010-09-28)
Yigang Teng, Qiuyu Yin, Ming ding, Fukun Zhao
RÉSUMÉ
In this study, we confirmed that at least three endo-β-1,4-glucanases existed in the digestive juice of the giant snail, Achatina fulica ferussac, by Congo red staining assay. One of these enzymes, a novel endo-β-1,4-glucanase (AfEG22), was purified 29.5-fold by gel filtration, anion exchange, and hydrophobic interaction chromatography. The carboxymethyl cellulose (CMC) hydrolytic activity of the purified enzyme was 12.3 U/mg protein. The molecular mass of AfEG22 was 22081 Da determined by MALDI-TOF. N-terminal amino acid sequencing revealed a sequence of EQRCTNQGGILKYYNT, which did not have significant homology with any proteins in BLAST database. The optimal pH and temperature for hydrolytic activity toward CMC were pH 4.0 and 50°C, respectively. AfEG22 was stable between pH 3.0 and pH 12.0 when incubated at 4°C for 3 h or at 37°C for 1 h. The enzyme remained more than 80% activity between pH 4.5 and pH 7.0 after incubation at 50°C for 1 h. AfEG22 possessed excellent thermostability as more than 70% activity was remained after incubation at 60°C for 3 h. Substrate specific analysis revealed that AfEG22 was a typical endo-β-1,4-glucanase. This is the first time to report a novel endo-β-1,4-glucanase with high stability from the digestive juice of A. fulica.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
β-Glucanase from Trichoderma longibrachiatum
Sigma-Aldrich
β-Glucanase from Aspergillus niger, powder, dark brown, ~1 U/mg