Documents
56701
Ferrule Upchurch sans bride
1/4-28 UNF ferrule for 1/8 in. O.D. tubing (requires Flangeless nut), ferrule only
Synonyme(s) :
Ferrules, 1/8in, Tefzel®, 10 pk
About This Item
Code UNSPSC :
41115700
Matériaux
Tefzel® ferrule
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
Upchurch Scientific, Inc P-300X
Raccords
1/4-28 UNF ferrule for 1/8 in. O.D. tubing (requires Flangeless nut)
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificats d'analyse (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Service Clients
Déjà en possession de ce produit ?
Les clients ont également consulté
Slide 1 of 3
1 of 3