Documents
22068
Raccords pour tubes, PTFE
PTFE front ferrule, Swagelok for 1/16 in. tubing
About This Item
Code UNSPSC :
41115717
Matériaux
PTFE front ferrule
Conditionnement
pkg of 5 ea
Raccords
Swagelok for 1/16 in. tubing
Description générale
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
Application
Swagelok® PTFE ferrules maybe used, for making all the connections between glass and PTFE present in the containment vessels, piping and sampling equipment in a bench scale biofilter system, in an experimental study investigating the effects of periods of non-use on biofilter performance.
Informations légales
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificats d'analyse (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Service Clients
Déjà en possession de ce produit ?
Les clients ont également consulté
Slide 1 of 1
1 of 1
Effect of periods of non-use on biofilter performance.
Jason MF and Loehr RC.
Journal of the Air & Waste Management Association (1995), 46 (6), 539-546 (1996)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)