Principaux documents
Z744249
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
for Bruker®, O.D. 10 mm, pack of 1 ea
Synonyme(s) :
EPR cryo dewar, EPR dewar
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.15
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Wilmad WG-816-B-Q
D.E.
10 mm
Description générale
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Informations légales
Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries